Small Intestine, Composite Slide
Small Intestine, Composite Slide, Section, Hematoxylin and Eosin
Catalog # 470176-962
Supplier: Avantor
Jump to:
Product Details & Documents
Investigate the digestive tract
- Identifying features clearly distinguishable
- Rigorous quality control standards
- Mammal
- Three regions (order from left to right): duodenum, jejunum, and ileum
Specifications
- Description:Small Intestine, Composite Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section