Slotted Weight Set
2×5 g, 1×10 g, 9×20 g, 1×50 g
Catalog # 470004-812
Supplier: PRECISION MOULDED POLYMERS LIMITED
Número de proveedor #: 1350806410000
Jump to:
Product Details & Documents
Marked Slotted Weights.
- Hook included
- 13 slotted weights included
All weights are machined stainless steel, accurately adjusted within B-grade tolerance, and marked.
Especificaciones
- No. of Weights:2×5 g, 1×10 g, 9×20 g, 1×50 g