Slab Easels
Supplier: JULE-ART INC.
Jump to:
About this item
Safely prop up flat specimens such as agate or fossils in a thin matrix. The smaller easel supports slabs up to 1/4" thick, the larger easel easily accommodates thicker slabs and crystal plates.
Slab Easel Size: 3"L x 2 1/2"W x 2 1/4"H. Package of four.
Thick Slab Easel Size: 2 1/2"L x 2 1/4"W x 2 3/4"H. Package of four.
1 Options Available Below
Product Details & Documents
Safely prop up flat specimens such as agate or fossils in a thin matrix. The smaller easel supports slabs up to 1/4" thick, the larger easel easily accommodates thicker slabs and crystal plates.
Slab Easel Size: 3"L x 2 1/2"W x 2 1/4"H. Package of four.
Thick Slab Easel Size: 2 1/2"L x 2 1/4"W x 2 3/4"H. Package of four.
Product Family Options
Product Information
- AvailabilityPrice
- 470018-778Slab Easels, Slab EaselSupplier Number: 36 A
- 470145-234Slab Easels, Thick Slab EaselSlab Easels, Thick Slab EaselDiscontinued-Supplier Number: 35