GPI Anatomicals® Skin Cancer Disk Set
470126-652
Small Business Enterprise
- Descripción:Skin Cancer Disk Set
- Altura:4 cm
- Longitud:10 cm
- No. of Keyed Items:5
- Referencia:470126-652
- Anchura:10 cm
- Nº de proveedor:3851
- No. of Parts:1
Depicts Five Cancerous Conditions
Teach students to visually identify and diagnose five types of cancerous skin conditions with this full-sized hinged model that presents enlarged views of each lesion. Includes clearly-labeled views of Dysplastic Nevus, Actinic Keratosis, Malignant Melanoma, Squamous Cell Carcinoma and Basal Cell Carcinoma that drive home the graphic and damaging effects of UV rays and sunburn.