ContactUsLinkComponent
Single-Use Fingerprint Powders
  470024-592
undefined
Single-Use Fingerprint Powders
  470024-592
 :  1-0301
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Descripción:
    Single-use fingerprint powder, black, ½ oz.
  • Referencia:
    470024-592
  • Nº de proveedor:
    1-0301
Ensure A Clean Sample Every Time
Guarantee that your latent print powder isn’t carrying any contaminants or DNA from crime scene to crime scene with these disposable single-use jars of Lightning® fingerprint powder. Because a new powder pack is used at each scene, there is no risk of cross-contamination, which can occur when the same tools are used from investigation to investigation. Each powder pack is sealed with a stiff foil lid, which keeps the powder clean and dry until you are ready to use it. Size: 1 oz. Available to U.S. customers only.