Orders LinkContactUsLinkComponent
Single Factor Water Test Kits
Single Factor Water Test Kits
Single Factor Water Test Kits
Certificates
Single Factor Water Test Kits
Single Factor Water Test Kits
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Simply Dip in Water and Read the Results
These kits feature a wide range of test factors for a fraction of the price of other single factor test kits. These test kits also eliminate the need for liquid reagents, hazardous chemicals, and glassware. Each kit comes with instructions.
Product Family Options
Información del producto
  • Availability
    Price
  • 470173-832
    Single factor water test kit, total chlorine, 30 tests
    Single factor water test kit, total chlorine, 30 tests
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $456.84
    $456.84
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $497.96
    $497.96
    Each (1UN)
       $456.84
    Case de 12 (1UN)
       $497.96
    Supplier Number: 481110
  • 470144-534
    Single factor water test kit, iron, 30 tests
    Single factor water test kit, iron, 30 tests
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $564.34
    $564.34
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $615.13
    $615.13
    Each (1UN)
       $564.34
    Case de 12 (1UN)
       $615.13
    Supplier Number: 480125
  • 470173-830
    Single factor water test kit, copper, 25 tests
    Single factor water test kit, copper, 25 tests
    Discontinued
    Supplier Number: 481111
  • 470173-840
    Single factor water test kit, pH, 30 tests
    Single factor water test kit, pH, 30 tests
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $309.04
    $309.04
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $336.85
    $336.85
    Each (1UN)
       $309.04
    Case de 12 (1UN)
       $336.85
    Supplier Number: 480104
  • 470173-834
    Single factor water test kit, nitrate/nitrite, 30 tests
    Single factor water test kit, nitrate/nitrite, 30 tests
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $541.90
    $541.90
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $590.68
    $590.68
    Each (1UN)
       $541.90
    Case de 12 (1UN)
       $590.68
    Supplier Number: 481109
  • 470144-634
    Single factor water test kit, total hardness, 30 tests
    Single factor water test kit, total hardness, 30 tests
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $309.04
    $309.04
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $336.85
    $336.85
    Each (1UN)
       $309.04
    Case de 12 (1UN)
       $336.85
    Supplier Number: 481108
  • 470144-546
    Single factor water test kit, Pesticides, 2 tests
    Single factor water test kit, Pesticides, 2 tests
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $806.16
    $806.16
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $878.72
    $878.72
    Each (1UN)
       $806.16
    Case de 12 (1UN)
       $878.72
    Supplier Number: 487996-SFK
  • 470173-838
    Single factor water test kit, heavy metals, 50 tests
    Single factor water test kit, heavy metals, 50 tests
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $577.77
    $577.77
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $629.76
    $629.76
    Each (1UN)
       $577.77
    Case de 12 (1UN)
       $629.76
    Supplier Number: 480309
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 