Single Eye Loupe
Single Lens Loupe
Catalog # 470105-654
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: MFL006
Jump to:
Product Details & Documents
These high quality loupes feature a 25mm glass lens mounted in a sturdy black plastic frame.
- Sturdy frame
- 25mm glass lens
Available in 6x magnification.
Especificaciones
- Magnification:6×
- Description:Single Lens Loupe