Orders LinkContactUsLinkComponent
Simple Squamous Epithelium Slides
Simple Squamous Epithelium Slides
Supplier:   Avantor
Simple Squamous Epithelium Slides
Supplier:  Avantor
Certificates
Simple Squamous Epithelium Slides
Simple Squamous Epithelium Slides
3 Options Available Below
Product Details & Documents
Single layer of epithelium that helps with diffusion and filtration.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Choose from several available stains and preparations for these tissue sections to get the view you're looking for. Slides are made from mammalian kidney tissue.
Hematoxylin and Eosin-stained slides show Bowman's Capsule, the thin section slide is plastic-embedded for greater viewing clarity.
Silver Nitrate-stained slides show Pavement tissue of mesentery, with cell borders in sharp relief.
Product Family Options
Información del producto
  • Orientation
    Stain
    Availability
    Price
  • 470182-600
    Simple Squamous Epithelium
    Simple Squamous Epithelium
    Section
    Hematoxylin and Eosin
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $193.60
    $193.60
    Each (1UN)
       $193.60
  • 470183-658
    Simple Squamous Epithelium
    Simple Squamous Epithelium
    Thin Section
    Hematoxylin and Eosin
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $357.95
    $357.95
    Each (1UN)
       $357.95
  • 470182-606
    Simple Squamous Epithelium
    Simple Squamous Epithelium
    Whole Mount
    Silver impreg.
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $185.00
    $185.00
    Each (1UN)
       $185.00