Orders LinkContactUsLinkComponent
Simple Cuboidal Epithelium Slides
Simple Cuboidal Epithelium Slides
Supplier:   Avantor
Simple Cuboidal Epithelium Slides
Supplier:  Avantor
Certificates
Simple Cuboidal Epithelium Slides
Simple Cuboidal Epithelium Slides
3 Options Available Below
Product Details & Documents
Aid in secretion and absorption of organs.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Choose from several available stains and preparations for these tissue sections to get the view you're looking for. Slides are made from mammal kidney, and demonstrate cuboidal cells in tubules. Thin section slide is plastic-embedded for greater viewing clarity.
Product Family Options
Información del producto
  • Orientation
    Stain
    Availability
    Price
  • 470182-610
    Simple Cuboidal Epithelium
    Simple Cuboidal Epithelium
    Section
    Hematoxylin and Eosin
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $193.60
    $193.60
    Each (1UN)
       $193.60
  • 470183-660
    Simple Cuboidal Epithelium
    Simple Cuboidal Epithelium
    Thin Section
    Hematoxylin and Eosin
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $234.25
    $234.25
    Each (1UN)
       $234.25
  • 470177-562
    Simple Cuboidal Epithelium
    Simple Cuboidal Epithelium
    Cross section
    Quadruple
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $193.60
    $193.60
    Each (1UN)
       $193.60