Shell Vials with Corks
Supplier: Avantor
Jump to:
Acerca de este artículo
These multipurpose glass vials come with tightly fitting cork plugs.
Packaging: Package of 12.
4 Options Available Below
Product Details & Documents
These multipurpose glass vials come with tightly fitting cork plugs.
Packaging: Package of 12.
Product Family Options
Información del producto
- Diameter ×HCap includedCap TypeAvailabilityPrice
- 470177-982Shell vials, 1/2 dramShell vials, 1/2 dram12×35 mmYesCork capEach (1UN)$40.43Paquete de 12 (1UN)$485.10
- 470177-984Shell vials, 1 dramShell vials, 1 dram15×45 mmYesCork capEach (1UN)$32.90Paquete de 12 (1UN)$394.80
- 470177-986Shell vials, 2 dramsShell vials, 2 drams17×60 mmYesCork capEach (1UN)$0.90Paquete de 12 (1UN)$10.80
- 470177-990Shell vials, 8 dramsShell vials, 8 drams25×95 mmYesCork capEach (1UN)$38.70Paquete de 12 (1UN)$464.40