Orders LinkContactUsLinkComponent
Shale (Carbonaceous)
Supplier:   Avantor
Shale (Carbonaceous)
Supplier:  Avantor
Certificates
Shale (Carbonaceous)
Shale (Carbonaceous)
3 Options Available Below
Product Details & Documents
Black, fissile; commonly with carbonized fossil plant impressions.
  • Collected in Pennsylvania
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kg of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Información del producto
  • Specimen Dimensions
    Availability
    Price
  • 470015-512
    Shale (Carbonaceous), Student Specimens
    Shale (Carbonaceous), Student Specimens
    1×1 - 1×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $227.38
    $227.38
    Paquete de 10
       $227.38
  • 470015-514
    Shale (Carbonaceous), Hand Specimen
    Shale (Carbonaceous), Hand Specimen
    2×3" - 3×4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $239.98
    $239.98
    Each (1UN)
       $239.98
  • 470026-268
    Shale (Carbonaceous), Bulk Pack, 1 kg
    Shale (Carbonaceous), Bulk Pack, 1 kg
    Size will vary
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $335.65
    $335.65
    Each (1UN)
       $335.65