Orders LinkContactUsLinkComponent
Ward's® Shale (Argillaceous)
Ward's® Shale (Argillaceous)
Supplier:   Avantor
Ward's® Shale (Argillaceous)
Supplier:  Avantor
Certificates
Ward's® Shale (Argillaceous)
Ward's® Shale (Argillaceous)
3 Options Available Below
Product Details & Documents
Typical green-gray, fissile, clay-rich.
  • Collected in New York
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Información del producto
  • Specimen Dimensions
    Availability
    Price
  • 470026-256
    Shale (Argillaceous), student specimens
    Shale (Argillaceous), student specimens
    1×1 - 1×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $227.95
    $227.95
    Paquete de 10
       $227.95
  • 470026-260
    Shale (Argillaceous), hand specimen
    Shale (Argillaceous), hand specimen
    2×3 - 3×4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $238.83
    $238.83
    Each (1UN)
       $238.83
  • 470026-258
    Shale (Argillaceous), bulk pack, 1 kg
    Shale (Argillaceous), bulk pack, 1 kg
    Size will vary
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $321.30
    $321.30
    Each (1UN)
       $321.30