Orders LinkContactUsLinkComponent
Ward's® Serpentine
Serpentine Thin Section
Catalog # 470024-930
Supplier:   Avantor
Ward's® Serpentine
Catalog # 470024-930
Supplier:  Avantor
Ward's® Serpentine
Ward's® Serpentine
Serpentine Thin Section
Price
$1,047.60
Log in for availability
Each (1UN)
Product Details & Documents
When minerals, rocks, and fossils cannot be accurately identified by macroscopic observation and testing, thin section slides can reveal the composition and structure of the specimens on a microscopic level.
Thin section slides are precisely ground to a thickness of 0.03 mm across the entire specimen area. Mineral and rock thin sections are mounted on 27 x 46 mm microscope slides, and protected by a glass coverslip. All thin section slides are labeled for reference.
Our excellent series of standard thin sections features important and interesting rock-forming mineral types most often found in field samples.
Specifications
  • Description:
    Serpentine Thin Section