Orders LinkContactUsLinkComponent
Separating a Mixture by Physical Properties Kit
Separating a Mixture by Physical Properties Kit
Separating a Mixture by Physical Properties Kit, 10
Catalog # 470039-664
Supplier:   Avantor
Separating a Mixture by Physical Properties Kit
Catalog # 470039-664
Supplier:  Avantor
Separating a Mixture by Physical Properties Kit
Separating a Mixture by Physical Properties Kit
Separating a Mixture by Physical Properties Kit, 10
Price
$4,300.80
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Physically Separate Different Substances
  • Two Levels of Instruction Included
  • Determine properties of different substances and collect data
Students identify physical properties of substances and then determine methods that can separate a mixture made of those substances. Methods include separation by properties including particle size, density, solubility, and magnetism. Directions for two levels of instruction are included. The elementary level activity stresses identification properties, use of tools, and using collected data appropriately in designing an experiment. The advanced level activity includes some mathematical analysis for understanding physical properties of matter. one 45-minute class period is required to complete the activity. Includes Instruction manual. A balance is required but not included.
Includes:
Magnetic wands
Strainers
Funnels
Aluminum pie plates
Filter Paper
Plastic Spoons
Pop beads
Salt
Sand
Steel nuts
Pebbles
Plastic Cups
Plastic Containers
Instructions
Required but Not Provided: Balance, Beakers, Water, Containers, Paper Towels.
Especificaciones
  • Description:
    Separating a Mixture by Physical Properties Kit
  • No. of Lab Groups:
    10
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 