Orders LinkContactUsLinkComponent
Self-Adhesive Lab Labels,
Self-Adhesive Lab Labels,
Self-adhesive lab labels
Catalog # 470150-028
Supplier:   EATON OFFICE SUPPLY
Supplier #:  AVE05434
Self-Adhesive Lab Labels,
Catalog # 470150-028
Self-Adhesive Lab Labels,
Self-Adhesive Lab Labels,
Self-adhesive lab labels
Price
$212.68
Log in for availability
Paquete de 1
Product Details & Documents
Specially made for laboratory use, these chemical resistant labels will adhere to just about any surface in your lab.
Size of each: 22×44 mm.
Packaging: Package of 500.
Specifications
  • Description:
    Self-adhesive lab labels