Screw Top Dropper Pipet Bottle
470012-952
- Capacidad:30 ml
- Referencia:470012-952
- Nº de proveedor:7-406-3-DZ
Molded of flint glass, screw tip with reusable, snap-on sealer tip.
- Glass pipet
- Closure included
- 30 ml capacity
These Boston round flint glass bottles include screw cap with dropper assembly are designed for general use. The glass is non-porous, which allows for easy cleaning or autoclaving.
: Pack of 12