Screw Lock Scalpel
Nickel Plated, 152 mm (6"), Screw Lock Scalpel, #22
Catalog # 470005-456
Supplier: DUNRITE INSTRUMENTS SE
Supplier #: 140705
Jump to:
Product Details & Documents
A versatile, inexpensive scalpel with a safety lock to hold the blade securely, yet allow for simple, safe changes.
The handle is fluted for a better grip, with a blade best suited for heavy dissections. The blade is held in place using a pin and screw spaced for blades fitting a number 4 scalpel.
Specifications
- Description:Screw Lock Scalpel
- Length:152 mm (6")
- Material:Nickel Plated
- Blade No.:#22