Screw Compressor Clamp with Closed Jaw
470144-828
- Descripción:Screw Compressor Clamp with Closed Jaw, Brass, Max. Jaw Opening, mm: 19
- Referencia:470144-828
- Nº de proveedor:HSCCL1
Used to control flow in flexible tubing.
- Fits tubes up to 19 mm
- Easy to use thumbscrew
This screw compressor clamps and holds flexible tubes securely closed.