Screw Compression Clamp with Open Jaw
Supplier: UNITED SCIENTIFIC SUPPLIES
Jump to:
Acerca de este artículo
Easily clamp tubes up to 1" in diameter.
- Zinc-plated brass
Can use as a closed clamp or open.
1 Options Available Below
Product Details & Documents
Easily clamp tubes up to 1" in diameter.
- Zinc-plated brass
Can use as a closed clamp or open.
Product Family Options
Información del producto
- AvailabilityPrice
- 470032-730Screw Compression Clamp with Open Jaw, Jaw Opening, mm: 13 to 19Screw Compression Clamp with Open Jaw, Jaw Opening, mm: 13 to 19Log in Para disponibilidadEach (1UN)$47.98Supplier Number: HSCOP1
- 470104-104Screw Compression Clamp with Open Jaw, Jaw Opening, mm: 16 to 25Screw Compression Clamp with Open Jaw, Jaw Opening, mm: 16 to 25Discontinued-Supplier Number: -