Sclerenchyma Slide
Sclerenchyma, Cross section, Quadruple
Catalog # 470182-528
Supplier: Avantor
Jump to:
Product Details & Documents
Heavily Stabilized Cell Walls
- Identifying features clearly distinguishable
- Rigorous quality control standards
Aristolochia. Provide structural support in plants.
Specifications
- Description:Sclerenchyma
- Stain:Quadruple
- Orientation:Cross section