Ward's® Scientific Method Poster
Scientific Method Poster, Unlaminated, 86 cm, 53 cm
Catalog # 470023-266
Supplier: Avantor
Jump to:
Product Details & Documents
Excellent Review Before Performing All Your Science Experiments
- Highlights each step of the Scientific Method
This excellent reference will reinforce to your students how scientists perform controlled experiments. It highlights each step of the Scientific Method including observing, questioning, forming a hypothesis, conducting an experiment, drawing a conclusion, and devising a theory. For each step discussed, it also provides examples from Francesco Redi’s famous experiment on Spontaneous Generation.
Especificaciones
- Width:53 cm
- Description:Scientific Method Poster
- Length:86 cm
- Finish:Unlaminated