Orders LinkContactUsLinkComponent
Miscellaneous Convenience Items
Miscellaneous Convenience Items
Wire, Insulated Copper 10 ft, 22 G, Red
Catalog # 470163-956
Supplier:   Wards
Número de proveedor #:  2806300
Miscellaneous Convenience Items
Catalog # 470163-956
Supplier:  Wards
Miscellaneous Convenience Items
Miscellaneous Convenience Items
Wire, Insulated Copper 10 ft, 22 G, Red
Price
$101.26
Log in Para disponibilidad
Paquete de 1
Product Details & Documents
Caution: WARNING: CHOKING HAZARD. Not for children under 3 years.
Especificaciones
  • Description:
    Wire, Insulated Copper 10 ft, 22 G, Red
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • Funnel, Poly, 114 mL (4 oz)
    Funnel, Poly, 114 mL (4 oz)
    470161-302
    Log in Para disponibilidad
    Each (1UN) - $83.30
Información del producto
  • Catalog #
    Availability
    Price
  • Single sheave
    Single sheave
    470049-642
    Log in Para disponibilidad
    Each (1UN) - $32.53
Información del producto
  • Catalog #
    Availability
    Price
  • Ball-Bearing Clamp Pulley
    Ball-Bearing Clamp Pulley
    470148-686
    Log in Para disponibilidad
    Each (1UN) - $262.80
Información del producto
  • Catalog #
    Availability
    Price
  • Weight, Hooked, 10 g to 1000 g, Cast Iron, Set/9
    Weight, Hooked, 10 g to 1000 g, Cast Iron, Set/9
    470003-584
    Log in Para disponibilidad
    Kit de 1 - $1,281.67
Información del producto
  • Catalog #
    Color
    Length
    Availability
    Price
  • Braided Nylon pulley cord, tensile strength 200 lbs. test
    Braided Nylon pulley cord, tensile strength 200 lbs. test
    470005-414
    White
    9 m (10 yds.)
    Log in Para disponibilidad
    Each (1UN) - $164.18
Información del producto
  • Catalog #
    Availability
    Price
  • Assorted Plastic Pulleys
    Assorted Plastic Pulleys
    470007-058
    Log in Para disponibilidad
    Each (1UN) - $430.68