Slime In the Bag Inquiry, Laboratory Activity
Product Details & Documents
In this fun activity, students are challenged to balance chemical equations, perform mole calculations, determine limiting reagents, and understand stoichiometry.
- Teacher and Student-Led activities
- Students practice stoichiometry
- For grades 6-12
- Material for 30
- Estimated lab time: 40 minutes
This activity uses chemistry to demonstrate how polymers form by combining two solutions. Students will try different amounts to create slime in a mess-proof way. The kit contains enough materials for 30 demonstrations. The activity requires 40 minutes.
Ordering information: This kit includes Polyvinyl alcohol, sodium borate solution, plastic bags, and instructions. Not included is food coloring and graduated cylinders.
Especificaciones
- Description:Slime in the bag inquiry, laboratory activity
- No. of Uses:30
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.