- Descripción:Biochemistry Teacher Set, 562 Pieces
- Referencia:470149-022
- Nº de proveedor:SET00611
Enhance Instructor-Led Biochemistry Lessons
Instructors can easily demonstrate key biochemistry concepts and chemical structure with this colorful molecular model. Featuring elements most commonly found in biochemistry, this set contains 270 color-coded spheres for carbon, oxygen, hydrogen, nitrogen, sulfur, phosphorus and metal, with holes for inserting rods that represent chemical bonds. Includes 200 short bonds, 30 medium bonds, and 30 long bonds, with link remover tool. Spheres range in diameter from 15mm to 22mm.
Contents:
Qty Element Color Holes Diameter
45 Carbon Black 4 22 mm
25 Carbon Black 3 22 mm
24 Oxygen Red 2 20 mm
10 Oxygen Red 1 15 mm
10 Oxygen Red 2-linear 20 mm
100 Hydrogen White 1 15 mm
10 Hydrogen White 2 15 mm
14 Nitrogen Blue 4 22 mm
12 Nitrogen Blue 3 22 mm
10 Nitrogen Blue 2 20 mm
2 Sulfur Yellow 2 22 mm
6 Phosphorus Purple 4 22 mm
1 Metal Grey 4 22 mm
1 Metal Grey 6 22 mm
30 Medium Bonds Grey
30 Long Bonds Grey
10 Medium Bonds Purple
200 Short Bonds White
Link Remover Tool