Orders LinkContactUsLinkComponent
Scheibler Desiccators
Scheibler Desiccators
Scheibler Desiccators
Scheibler Desiccators
Made of clear glass with thick uniform walls and a heavy, broad flange. Cover with molded knob is ground to fit the flange perfectly. Porcelain plates sold separately.
Product Family Options
  • Description
  • Scheibler Desiccators, ID, mm: 150
    Each (1UN)
    Each (1UN) - $2,529.13
    $2,529.13
    Each (1UN)
       $2,529.13
  • Scheibler Desiccators, ID, mm: 250