Orders LinkContactUsLinkComponent
Scenedesmus Slide
Scenedesmus Slide
Scenedesmus, Whole mount, Fast Green and Mallory's
Catalog # 470180-982
Supplier:   Avantor
Scenedesmus Slide
Catalog # 470180-982
Supplier:  Avantor
Scenedesmus Slide
Scenedesmus Slide
Scenedesmus, Whole mount, Fast Green and Mallory's
Price
$135.18
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Genus of Green Algae
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Colonies of elongated cells united laterally; the terminal cells have spine-like processes.
Especificaciones
  • Description:
    Scenedesmus
  • Stain:
    Fast Green and Mallory's
  • Orientation:
    Whole mount