Sarcocystis Slide
Sarcocystis, Section, Iron Hematoxylin
Catalog # 470181-966
Supplier: Avantor
Jump to:
Product Details & Documents
Asexual Multiplication within the sarcocyst Occurs
- Identifying features clearly distinguishable
- Rigorous quality control standards
Striated muscle shows thread-like sarcocystis, historically known as Miescher’s tubules.
Especificaciones
- Description:Sarcocystis
- Stain:Iron Hematoxylin
- Orientation:Section