Orders LinkContactUsLinkComponent
Ward's® Sandstone (White)
Supplier:   Avantor
Ward's® Sandstone (White)
Supplier:  Avantor
Certificates
Ward's® Sandstone (White)
Ward's® Sandstone (White)
3 Options Available Below
Product Details & Documents
Well-cemented, medium-grained orthoquartzite.
  • Collected in West Virginia
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Información del producto
  • Specimen Dimensions
    Availability
    Price
  • 470025-842
    Sandstone (White), student specimens
    Sandstone (White), student specimens
    1×1 - 1×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $225.10
    $225.10
    Paquete de 10
       $225.10
  • 470025-846
    Sandstone (White), hand specimen
    Sandstone (White), hand specimen
    2×2" - 3×4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $282.93
    $282.93
    Each (1UN)
       $282.93
  • 470025-844
    Sandstone (White), bulk pack, 1 kg
    Sandstone (White), bulk pack, 1 kg
    Size will vary
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $329.35
    $329.35
    Each (1UN)
       $329.35