Orders LinkContactUsLinkComponent
Ward's® Sandstone (Siliceous)
Ward's® Sandstone (Siliceous)
Supplier:   Avantor
Ward's® Sandstone (Siliceous)
Supplier:  Avantor
Certificates
Ward's® Sandstone (Siliceous)
Ward's® Sandstone (Siliceous)
4 Options Available Below
Product Details & Documents
Gray, fine to medium-grained “Berea grit”.
  • Collected in Ohio
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Información del producto
  • Specimen Dimensions
    Availability
    Price
  • 470026-198
    Sandstone (Siliceous), student specimens
    Sandstone (Siliceous), student specimens
    1×1 - 1×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $202.18
    $202.18
    Paquete de 10
       $202.18
  • 470026-202
    Sandstone (Siliceous), hand specimen
    Sandstone (Siliceous), hand specimen
    2×2 - 3×4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $227.38
    $227.38
    Each (1UN)
       $227.38
  • 470026-200
    Sandstone (Siliceous), bulk pack, 1 kg
    Sandstone (Siliceous), bulk pack, 1 kg
    Size will vary
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $296.10
    $296.10
    Each (1UN)
       $296.10
  • 470025-052
    Sandstone (Siliceous) thin section
    Sandstone (Siliceous) thin section
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $44.99
    $44.99
    Each (1UN)
       $44.99