Sandpaper
Sandpaper, Medium
Catalog # 470147-480
Supplier: SAINT GOBAIN ABRASIVES
Número de proveedor #: 07660747735
Jump to:
Product Details & Documents
Medium sandpaper in 9" x 11" sheets.
- Medium grit
- Multipurpose
- 9" x 11" sheets
In 9" x 11" sheets.
Especificaciones
- Description:Sandpaper, Medium