Salmonella enteritidis Slide
Salmonella enteritidis, Smear, Gram (-)
Catalog # 470180-916
Supplier: Avantor
Jump to:
Product Details & Documents
Facultative Anerobe
- Identifying features clearly distinguishable
- Rigorous quality control standards
Rods. Human intestinal pathogen.
Especificaciones
- Description:Salmonella enteritidis
- Stain:Gram (-)
- Orientation:Smear