Rubber Policemen with Glass Rods
Catálogo # 470123-286
Proveedor: UNITED SCIENTIFIC SUPPLIES
Especificaciones
- Ø:5 mm
- Descripción:Rubber policemen with glass rods
- Longitud:6"
- Referencia:470123-286
- Nº de proveedor:GRPL06
Especificaciones
Acerca de este artículo
Rubber policeman attached to glass rod facilitates scraping and mixing.
- Soft flint glass
- Fire polished ends
Firm, flexible gum rubber tubing with cemented diagonal end on a 15 cm glass rod. Size: 3mm dia. Package of 12.