Round Trivet
Round Trivet, 18 cm
Catalog # 470157-568
Supplier: HUTZLER MFG CO INC SE
Número de proveedor #: 504-RED
Jump to:
Product Details & Documents
Affordable and practical, these melamine trivets are heat-resistant and remain cool to the touch. Colors may vary.
Affordable and practical, these melamine trivets are heat-resistant and remain cool to the touch. Colors may vary. Size: 18cm dia.
Especificaciones
- Ext.Ø:18 cm
- Description:Round Trivet