PYREX® Narrow Mouth Reagent Bottles, Corning Life Sciences
PYREX® borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These narrow mouth reagent storage bottles have sturdy ground necks with standard taper stoppers. Replacement stoppers are available.
Frequently Bought Together