Orders LinkContactUsLinkComponent
Round Carboys
Round Carboys
Round Carboys
Certificates
Round Carboys
3 Options Available Below
Product Details & Documents
Suitable for collecting and dispensing distilled water, reagents, and acids, these low-density polyethylene carboys are graduated in 1 gallon and 5 L increments.
  • Two larger sizes also have convenient wide shoulder handles
The polypropylene spigot is molded in one piece with the bottle for added strength and leak resistance.
Product Family Options
Información del producto
  • Availability
    Price
  • 470211-768
    Round Carboy, 4 L
    Round Carboy, 4 L
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,590.00
    $3,590.00
    Log in Para disponibilidad
    Case de 6 (1UN)
    Case de 6 (1UN) - $4,183.94
    $4,183.94
    Each (1UN)
       $3,590.00
    Case de 6 (1UN)
       $4,183.94
    Supplier Number: 2318-0010
  • 470211-770
    Round Carboy, 10 L
    Round Carboy, 10 L
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $5,160.00
    $5,160.00
    Log in Para disponibilidad
    Case de 6 (1UN)
    Case de 6 (1UN) - $6,019.29
    $6,019.29
    Each (1UN)
       $5,160.00
    Case de 6 (1UN)
       $6,019.29
    Supplier Number: 2318-0020
  • 470211-772
    Round Carboy, 20 L
    Round Carboy, 20 L
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $6,170.00
    $6,170.00
    Log in Para disponibilidad
    Case de 4 (1UN)
    Case de 4 (1UN) - $7,198.56
    $7,198.56
    Each (1UN)
       $6,170.00
    Case de 4 (1UN)
       $7,198.56
    Supplier Number: 2318-0050