Rock and Mineral Test Kit
Catálogo # 470018-648
Proveedor: Avantor
Especificaciones
- Descripción:Rock and Mineral Test Kit
- Referencia:470018-648
- Nº de proveedor:WARD470018-648
Especificaciones
Acerca de este artículo
Comprehensive, Economical Test Kit.
- Economical
- Perform tests on rocks and minerals
A handy zippered pouch contains all the equipment you need to perform a variety of physical properties tests on rocks and minerals.
Información de pedido: This kit includes a streak plate for streak color; glass plate, nail, and copper strip for hardness; magnet for iron content; magnifier; and acid bottle (acid not included) for CaCO3 test.