Right Angle Prism
Prism, Right Angle, 32 mm x 50 mm
Catalog # 470104-440
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: PAR048
Jump to:
Product Details & Documents
A must for your Optics lab.
- Clear acrylic
- Demonstrates refraction and internal reflection
A polished acrylic glass prism measures 48 x 50 mm in length.
Especificaciones
- Description:Prism, Right Angle, 32 mm x 50 mm