Orders LinkContactUsLinkComponent
Right Angle Clamp Holders
Right Angle Clamp Holders
Supplier:   HUMBOLDT MFG CO
Right Angle Clamp Holders
Supplier:  HUMBOLDT MFG CO
Certificates
Right Angle Clamp Holders
Right Angle Clamp Holders
2 Options Available Below
Product Details & Documents
The perpendicular V-shaped jaws of these clamp holders are desiged to support rods and clamp extensions at a 90° angle.
Holders feature thumbscrews at a 180° angle.
Product Family Options
Información del producto
  • Availability
    Price
  • 470019-546
    Right Angle Clamp Holder, for 1/2" dia. Rods
    Right Angle Clamp Holder, for 1/2" dia. Rods
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $298.49
    $298.49
    Each (1UN)
       $298.49
    Supplier Number: H-8300
  • 470019-548
    Right Angle Clamp Holder, for 3/4" dia. Rods
    Right Angle Clamp Holder, for 3/4" dia. Rods
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $327.81
    $327.81
    Each (1UN)
       $327.81
    Supplier Number: H-8320