Rheostat
Supplier: VWR International
Jump to:
Acerca de este artículo
This newly designed rheostat features a protective cover.
- Oxidized Resistance Wire
- Safety-Socket Terminals
- Resistance: ±10%
- Protective Cover
It's wound with a heavily oxidized resistance wire on a tube and fitted with 4 mm safety-socket terminals.
Its resistance is ±10% compared to nominal value.
4 Options Available Below
Product Details & Documents
This newly designed rheostat features a protective cover.
- Oxidized Resistance Wire
- Safety-Socket Terminals
- Resistance: ±10%
- Protective Cover
It's wound with a heavily oxidized resistance wire on a tube and fitted with 4 mm safety-socket terminals.
Its resistance is ±10% compared to nominal value.
Product Family Options
Información del producto
- AvailabilityPrice
- 470003-094Covered Rheostat, 10 Ohm, 4A
- 470152-306Covered Rheostat, 100 Ohm 1.75 A
- 470003-096Covered Rheostat, 330 Ohm, 0.95 A
- 470003-098Covered Rheostat, 1000 Ohm 0.5 A