PYREX® Reusable Mohr-Type Pipettes
25 ml, Pyrex® reusable mohr-type pipettes
Supplier: CORNING CA
Número de proveedor #: 7065-25
Jump to:
Product Details & Documents
These reusable 1 ml PYREX® glass pipettes are color-coded by size for easy identification and sorting.
- Tips are ground, beveled, and tempered
- Colored graduations are enameled onto the glass
- Accuracy to ±0.01 ml
Amber graduations are fused into glass so they won’t wear off. The graduation intervals on these pipettes are 0.1 ml. They are calibrated 'to deliver' (TD) their total capacity without blow-out.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Capacity:25 ml
- Description:Pyrex® reusable mohr-type pipettes