Resistance Blocks
Supplier: VWR International
Jump to:
About this item
Economical yet Sturdy
- Simple to use
- Safe to handle
Students can use these resistance units to construct various types of circuits. Built with a high stability metal film element, these durable resistance units are more economical that resistance coils. Accuracy: 1%. 1W.
6 Options Available Below
Product Details & Documents
Economical yet Sturdy
- Simple to use
- Safe to handle
Students can use these resistance units to construct various types of circuits. Built with a high stability metal film element, these durable resistance units are more economical that resistance coils. Accuracy: 1%. 1W.
Product Family Options
Product Information
- AvailabilityPrice
- 470121-0905 Ohm Resistance block
- 470121-09210 Ohm Resistance block
- 470121-09420 Ohm Resistance block
- 470121-09650 Ohm Resistance block
- 470121-098100 Ohm Resistance block
- 470121-100200 Ohm Resistance block