Replacement Pump Oil
Replacement Oil for Vacuum Pumps, 946 mL
Catalog # 470104-444
Supplier: FISCHER TECHNICAL CO SE
Número de proveedor #: OIL-012
Jump to:
Product Details & Documents
This is a specially prepared, low vapor pressure mineral oil. It is rust-inhibiting and foam resistant, and permits water condensates to drop out of suspension.
Especificaciones
- Description:Replacement Oil for Vacuum Pumps, 946 mL