Ward's® Refractive Index Immersion Slide Set
Refractive Index Immersion Slide Set, 10
Catalog # 470024-872
Supplier: Avantor
Jump to:
Product Details & Documents
Ten New Slides in a Complete Teaching Set
- Hand Made by Wards® staff Geologists
- 10 Specially Selected Samples
- Demonstrates Refractive Index of Minerals
Students learn to evaluate refractive index using the Becke Line method.
Set includes teacher’s guide and student worksheets.
10 slides with one mineral each: fluorite, microcline, albite, quartz, labradorite, biotite, calcite, hornblende, almandine, granite.
Especificaciones
- Description:Refractive Index Immersion Slide Set
- No. of Slides:10