Red Liquid Filled Wall Thermometer
Thermometer, Taylor Instruments, −20...+120 °F / −30...+50 °C , Red Liquid
Catalog # 470003-334
Supplier: TAYLOR PRECISION PRODUCTS SE
Número de proveedor #: 5154
Jump to:
Product Details & Documents
Vivid red fluid makes this one easy-to-read thermometer.
Suitable for use indoors or out. Overall dimensions are 19 cm x 6 cm.
Especificaciones
- Description:Thermometer, Taylor Instruments
- Fill Solution:Red Liquid
- Range:−20...+120 °F / −30...+50 °C