Recto-Anal Junction Slide
Recto-Anal Junction, Longitudinal section, Hematoxylin and Eosin
Catalog # 470182-882
Supplier: Avantor
Jump to:
Product Details & Documents
Digestive System Structure
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Region of transition from glandular to stratified squamous epithelium.
Especificaciones
- Description:Recto-Anal Junction
- Stain:Hematoxylin and Eosin
- Orientation:Longitudinal section