Orders LinkContactUsLinkComponent
Rectangular Supports
Rectangular Supports
Supplier:   SHIVDIAL SUD & SONS
Rectangular Supports
Certificates
Rectangular Supports
Rectangular Supports
2 Options Available Below
Product Details & Documents
Tip-free stability for experiments.
  • Glassware can sit directly on base
  • Rods are zinc-plated steel
Heavy cast iron bases with black enamel finish. Includes a removable zinc-plated steel rod.
Product Family Options
Información del producto
  • Base Dimensions
    Rod Diameter
    Rod Length
    Availability
    Price
  • 4×6"
    ⁵/₁₆"
    18"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $388.56
    $388.56
    Each (1UN)
       $388.56
    Supplier Number: SSB4X6-CI
  • 5×8"
    ⅜"
    20"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $287.65
    $287.65
    Log in Para disponibilidad
    Case de 6 (1UN)
    Case de 6 (1UN) - $1,320.70
    $1,320.70
    Each (1UN)
       $287.65
    Case de 6 (1UN)
       $1,320.70
    Supplier Number: CH12321/2