Orders LinkContactUsLinkComponent
Rectangular Glass Coverslips
Rectangular Glass Coverslips
Rectangular Glass Coverslips
Certificates
Rectangular Glass Coverslips
Rectangular Glass Coverslips
3 Options Available Below
Product Details & Documents
Traditional rectangular coverslip style
  • Quality slide making supplies
  • Traditional coverslip type
Traditional style coverslips to use when making slides in the laboratory.
#1 Thickness: 0.15 mm thick. Approximately 75–100 coverslips per ounce.
#2 Thickness: 0.2 mm thick. Approximately 50-75 coverslips per ounce.
Product Family Options
Información del producto
  • L×W
    Thickness
    Numbering
    Availability
    Price
  • 470019-008
    Glass cover slips, rectangular
    Glass cover slips, rectangular
    40×22 mm
    0.15 mm
    #1
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 1
    Paquete de 1 - $87.42
    $87.42
    Paquete de 1
       $87.42
    Supplier Number: 94-2240
  • 470019-010
    Glass cover slips, rectangular
    Glass cover slips, rectangular
    40×24 mm
    0.15 mm
    #1
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 1
    Paquete de 1 - $87.42
    $87.42
    Paquete de 1
       $87.42
    Supplier Number: 94-2440
  • 470145-746
    Glass cover slips, rectangular
    Glass cover slips, rectangular
    40×22 mm
    0.2 mm
    #2
    Log in Para disponibilidad
    Log in Para disponibilidad
    Onza
    Onza - $87.42
    $87.42
    Onza
       $87.42
    Supplier Number: 94-22402