Orders LinkContactUsLinkComponent
Rectangular Bases
Rectangular Bases
Supplier:   VWR International
Rectangular Bases
Supplier:  VWR International
Certificates
Rectangular Bases
Rectangular Bases
3 Options Available Below
Product Details & Documents
Essential for Any Chemistry or Physics Laboratory
  • 3 mm thick steel
  • Baked-on enamel finish
  • 2 tapped hole positions
One hole is in the base top at the center of the shorter side for standard ring-stand use and one other at the center of the longer side for titrations and specific gravity determinations.
Ordering information: Rod sold separately.
Product Family Options
Información del producto
  • Base Dimensions
    Height
    Thread Size
    Availability
    Price
  • 470016-124
    Base, rectangular, red
    Base, rectangular, red
    5.5×9"
    23 mm
    1/2 - 13
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $226.52
    $226.52
    Each (1UN)
       $226.52
  • 470123-470
    Base, rectangular, red
    Base, rectangular, red
    4×6"
    23 mm
    5/6 - 18
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $191.88
    $191.88
    Each (1UN)
       $191.88
  • 470123-474
    Base, rectangular, red
    Base, rectangular, red
    6×11"
    23 mm
    1/2 - 13
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $325.17
    $325.17
    Log in Para disponibilidad
    Case de 10 (1UN)
    Case de 10 (1UN) - $354.43
    $354.43
    Each (1UN)
       $325.17
    Case de 10 (1UN)
       $354.43