Eisco® Quiet Air Source
Quiet air source, 120 V
Catalog # 470199-916
Supplier: VWR International
Jump to:
Product Details & Documents
The Quiet Air Source system powers air tracks and air tables with minimal noise.
- Economical
- Compact
- Quiet Air Delivery
- Air Hose Included
- Source Powers Air Tracks and Tables
Quieter than other air sources yet powerful enough to easily float fully-loaded gliders, this compact and economical air source is a technical breakthrough.
In operation, the sound level does not exceed that of quiet conversation.
Ordering information: This kit includes a 1.5 m long, 3 cm dia. air hose and a 4 cm hose adapter to fit other Air Tracks and Tables.
Especificaciones
- Description:Quiet air source, 120 V