Orders LinkContactUsLinkComponent
Ward's® Quartzite (Green)
Supplier:   Avantor
Ward's® Quartzite (Green)
Supplier:  Avantor
Certificates
Ward's® Quartzite (Green)
Ward's® Quartzite (Green)
3 Options Available Below
Product Details & Documents
Chlorite-bearing.
  • Collected in Ontario, Canada
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Información del producto
  • Specimen Dimensions
    Availability
    Price
  • 470025-848
    Quartzite (Green), student specimens
    Quartzite (Green), student specimens
    1×1 - 1×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $332.20
    $332.20
    Paquete de 10
       $332.20
  • 470025-850
    Quartzite (Green), bulk pack, 1 kg
    Quartzite (Green), bulk pack, 1 kg
    Size will vary
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $529.78
    $529.78
    Each (1UN)
       $529.78
  • 470025-852
    Quartzite (Green), hand specimen, Each
    Quartzite (Green), hand specimen, Each
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $444.45
    $444.45
    Each (1UN)
       $444.45